Merck
In vivo toxicity assessment of non-cadmium quantum dots in BALB/c mice.
Nanomedicine : nanotechnology, biology, and medicine (2014-12-03)
Guimiao Lin, Qingling Ouyang, Rui Hu, Zhangchi Ding, Jinglin Tian, Feng Yin, Gaixia Xu, Qiang Chen, Xiaomei Wang, Ken-Tye Yong
ABSTRACT
Along with widespread usage of QDs in electronic and biomedical industries, the likelihood of QDs exposure to the environment and humans is deemed to occur when the QD products are degraded or handled as waste for processing. To date, there are very few toxicological reports available in the literature for non-cadmium QDs in animal models. In this work, we studied the long term in vivo toxicity of InP/ZnS QDs in BALB/c mice. The biodistribution, body weight, hematology, blood biochemistry, and organ histology were determined at a very high dosage (25 mg/kg) of InP/ZnS QDs over 84 days period. Our results manifested that the QDs formulation did not result in observable toxicity in vivo within the evaluation period, thereby suggesting that the InP/ZnS QDs can be utilized as optical probes or nanocarrier for selected in vivo biological applications when an optimized dosage is employed. This study investigated the toxicity of quantum dots in BALB/c mice, and concluded that no organotoxicity was detectable despite of using high concentration of InP/ZnS quantum dots with prolonged exposure of 3 months.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, ACS reagent, 70%