Merck
  • Oral supplements of aqueous extract of tomato seeds alleviate motor abnormality, oxidative impairments and neurotoxicity induced by rotenone in mice: relevance to Parkinson's disease.
Oral supplements of aqueous extract of tomato seeds alleviate motor abnormality, oxidative impairments and neurotoxicity induced by rotenone in mice: relevance to Parkinson's disease.
Neurochemical research (2014-05-17)
Krishna Gokul, Muralidhara
ABSTRACT
Although tomato seeds (an industrial by-product) are known to contain several bioactive compounds, studies describing their health effects are limited. Previously, we evidenced that aqueous extract of tomato seeds (TSE) markedly attenuated rotenone (ROT)-induced oxidative stress and neurotoxicity in Drosophila system. This study investigated the neuroprotective effect of TSE in a chronic ROT model of neurotoxicity in mice. Initially, we assessed the potential of oral supplements of TSE to modulate the levels of endogenous markers of oxidative stress in brain regions of mice. Subsequently, employing a co-exposure paradigm, the propensity of TSE (100 mg/kg bw, 3 weeks) to attenuate ROT-induced behavioral phenotype (gait abnormalities, anxiety-like state), oxidative dysfunctions and neurotoxicity was examined. We found that mice provided with TSE supplements exhibited progressive improvement in gait pattern and exploratory behavior. TSE markedly offset ROT-induced oxidative impairments, restored reduced glutathione levels, antioxidant defenses (superoxide dismutase, glutathione peroxidase) and protein carbonyls content in brain regions. Specifically, TSE effectively diminished ROT induced elevation in the activity levels of acetylcholinesterase and restored the dopamine levels in striatum. Interestingly, in mitochondria, TSE was able to restore the activity of mitochondrial complexes and redox state. Collectively, our findings in the chronic ROT model demonstrate the ability of TSE to alleviate behavioral phenotype, oxidative stress, mitochondrial dysfunction and neurotoxicity. Further studies in dopaminergic cell models are necessary to understand the precise molecular mechanism/s by which tomato seed bioactives offer significant neuroprotection.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C