Merck
  • BET bromodomain inhibitors block growth of pancreatic cancer cells in three-dimensional collagen.
BET bromodomain inhibitors block growth of pancreatic cancer cells in three-dimensional collagen.
Molecular cancer therapeutics (2014-05-09)
Vaibhav Sahai, Krishan Kumar, Lawrence M Knab, Christina R Chow, Sania S Raza, David J Bentrem, Kazumi Ebine, Hidayatullah G Munshi
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) is associated with pronounced fibrosis that contributes to chemoresistance, in part, through increased histone acetylation. Because bromodomain (BRD) and extra terminal domain (BET) proteins are "readers" of histone acetylation marks, we targeted BET proteins in PDAC cells grown in three-dimensional collagen. We show that treatment with BET inhibitors decreases growth of PDAC cells (AsPC1, CD18, and Panc1) in collagen. Transfection with siRNA against BRD4, which is increased in human PDAC tumors, also decreases growth of PDAC cells. BET inhibitors additionally decrease growth in collagen of PDAC cells that have undergone epithelial-to-mesenchymal transition or have become resistant to chemotherapy. Although BET inhibitors and BRD4 siRNA repress c-MYC only in AsPC1 and CD18 cells, downregulating c-MYC decreases growth of all three PDAC cell lines in collagen. FOSL1, which is also targeted by BET inhibitors and BRD4 siRNA in AsPC1, CD18, and Panc1 cells, additionally regulates growth of all three PDAC cell lines in collagen. BET inhibitors and BRD4 siRNA repress HMGA2, an architectural protein that modulates chromatin state and also contributes to chemoresistance, in PDAC cells grown in collagen. Importantly, we show that there is a statistically significant correlation between BRD4 and HMGA2 in human PDAC tumors. Significantly, overexpression of HMGA2 partially mitigates the effect of BET inhibitors on growth and c-MYC and/or FOSL1 expression in collagen. Overall, these results demonstrate that BET inhibitors block growth of PDAC cells in collagen and that BET proteins may be potential targets for the treatment of pancreatic cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Brd4
Sigma-Aldrich
Chromatin Immunoprecipitation (ChIP) Assay Kit, Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads.
Sigma-Aldrich
MISSION® esiRNA, targeting human BRD4
Sigma-Aldrich
MISSION® esiRNA, targeting human FOSL1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fosl1