Merck
  • Generation of induced pluripotent stem cells derived from primary and secondary myelofibrosis patient samples.
Generation of induced pluripotent stem cells derived from primary and secondary myelofibrosis patient samples.
Experimental hematology (2014-05-27)
Masataka Hosoi, Keiki Kumano, Kazuki Taoka, Shunya Arai, Keisuke Kataoka, Koki Ueda, Yasuhiko Kamikubo, Naoya Takayama, Makoto Otsu, Koji Eto, Hiromitsu Nakauchi, Mineo Kurokawa
ABSTRACT
Induced pluripotent stem cells (iPS) derived from disease cells are expected to provide a new experimental material, especially for diseases from which samples are difficult to obtain. In this study, we generated iPS from samples from patients with primary and secondary myelofibrosis. The primary myelofibrosis cells had chromosome 13q deletions, and the secondary myelofibrosis (SMF) cells had JAK2V617F mutations. The myelofibrosis patient cell-derived iPS (MF-iPS) were confirmed as possessing these parental disease-specific genomic markers. The capacity to form three germ layers was confirmed by teratoma assay. By co-culture with specific feeder cells and cytokines, MF-iPS can re-differentiate into blood progenitor cells and finally into megakaryocytes. We found that mRNA levels of interleukin-8, one of the candidate cytokines related to the pathogenesis of myelofibrosis, was elevated predominantly in megakaryocytes derived from MF-iPS. Because megakaryocytes from myelofibrosis clones are considered to produce critical mediators to proliferate fibroblasts in the bone marrow and iPS can provide differentiated cells abundantly, the disease-specific iPS we established should be a good research tool for this intractable disease.
Product Description
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
IL-3 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
VEGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Trisodium citrate dihydrate, meets USP testing specifications
Sigma-Aldrich
Interleukin-3 human, IL-3, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
IL-3 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture