Merck
  • Curcumin enhances cell-surface LDLR level and promotes LDL uptake through downregulation of PCSK9 gene expression in HepG2 cells.
Curcumin enhances cell-surface LDLR level and promotes LDL uptake through downregulation of PCSK9 gene expression in HepG2 cells.
Molecular nutrition & food research (2014-08-29)
Mi-Hsueh Tai, Po-Kong Chen, Pei-Yi Chen, Ming-Jiuan Wu, Chi-Tang Ho, Jui-Hung Yen
ABSTRACT
Curcumin has been demonstrated as having numerous desirable characteristics, such as antioxidant, anti-inflammatory, and antiatherogenic activities. We report the hypocholesterolemic effect and molecular mechanism of curcumin. We found that curcumin enhanced LDL receptor (LDLR) level on the cell surface, as well as LDLR activity; however, LDLR transcription and mRNA stability were not affected. Furthermore, we found that proprotein convertase subtilisin/kexin type 9 (PCSK9) gene was downregulated at the transcriptional level by curcumin, leading to an increase in LDL uptake in HepG2 cells. The curcumin-responsive element of the PCSK9 promoter, a binding site for hepatocyte nuclear factor 1α (HNF-1α), was also identified. We demonstrated that curcumin reduced the nuclear abundance of hepatocyte nuclear factor 1α, resulting in its attenuated interaction with the PCSK9 promoter and leading to a downregulation of PCSK9 expression. Finally, we showed that curcumin decreased the statin-induced PCSK9 expression and potentially synergized with statin administration. Current results indicate that curcumin suppression of PCSK9 expression is associated with increases in cell-surface LDLR and LDLR activity in hepatic cells and it acts in a molecular mechanism that is distinct from the statins. Curcumin exhibits hypolipidemic activity and may serve as a useful supplement to statin treatment for hypercholesterolemia.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Supelco
Lovastatin, Pharmaceutical Secondary Standard; Certified Reference Material
Lovastatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Curcumin, primary reference standard
Supelco
Curcumin, matrix substance for MALDI-MS, ≥99.5% (HPLC)
Supelco
Curcumin, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard