Merck
  • Effects of remote ischemic preconditioning and myocardial ischemia on microRNA-1 expression in the rat heart in vivo.
Effects of remote ischemic preconditioning and myocardial ischemia on microRNA-1 expression in the rat heart in vivo.
Shock (Augusta, Ga.) (2014-07-01)
Timo Brandenburger, Hilbert Grievink, Nicole Heinen, Franziska Barthel, Ragnar Huhn, Friederike Stachuletz, Malte Kohns, Benedikt Pannen, Inge Bauer
ABSTRACT
Remote ischemic preconditioning (RIPC) is an easily applicable method for protecting the heart against a subsequent ischemia and reperfusion (I/R) injury. However, the exact molecular mechanisms underlying RIPC are unknown. We examined the involvement of microRNAs (miRNAs) and in particular the expression of miRNA-1 (miR-1) in RIPC and myocardial ischemia. Remote ischemic preconditioning was conducted by four cycles of 5-min bilateral hind-limb ischemia in male Wistar rats. Cardiac ischemia was induced by ligation of the left anterior descending coronary artery for 35 min followed by 2 or 6 h of reperfusion. MicroRNA expression was analyzed by Taqman miRNA arrays and quantitative polymerase chain reaction assays. Luciferase assays were performed to validate the miR-1 target gene brain-derived neurotrophic factor (BDNF). Brain-derived neurotrophic factor mRNA and protein levels were analyzed by quantitative polymerase chain reaction and enzyme-linked immunosorbent assay. Remote ischemic preconditioning led to a differential expression of miRNAs. The most abundant cardiac miRNA, miR-1, was downregulated by RIPC without following ischemia as well as after I/R and RIPC followed by I/R after 2 h of reperfusion. After 6 h of reperfusion, RIPC led to an upregulation of miR-1, whereas ischemia had no effect on miR-1 expression. Luciferase assays confirmed the interaction of miR-1 with BDNF, a protein that has been shown to exert cardioprotective effects. Brain-derived neurotrophic factor protein levels in rat hearts measured by enzyme-linked immunosorbent assay were not significantly altered after 2 or 6 h of reperfusion in all intervention groups. Remote ischemic preconditioning leads to changes in the expression levels of the most abundant cardiac miRNA, miR-1. MicroRNA 1 levels did not correlate with protein levels of BDNF, a known miR-1 target, in vivo. Further studies are needed to explore the biological significance of changes in miR-1 expression levels and the potential interaction with BDNF in RIPC-induced cardioprotection.
Sigma-Aldrich
α-Tri-Calcium phosphate, Reagent for transient & stable DNA transfections
Pricing and availability is not currently available.
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (unsintered powder)
Pricing and availability is not currently available.
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (sintered Powder)
Pricing and availability is not currently available.