Merck
  • Pharmacokinetics of dolutegravir when administered with mineral supplements in healthy adult subjects.
Pharmacokinetics of dolutegravir when administered with mineral supplements in healthy adult subjects.
Journal of clinical pharmacology (2014-12-03)
Ivy Song, Julie Borland, Niki Arya, Brian Wynne, Stephen Piscitelli
ABSTRACT
All commercially available integrase inhibitors are 2-metal binders and may be affected by co-administration with metal cations. The purpose of this study was to evaluate the effect of calcium and iron supplements on dolutegravir pharmacokinetics and strategies (dose separation and food) to attenuate the effects if significant reductions in dolutegravir exposure were observed. This was an open-label, crossover study that randomized 24 healthy subjects into 1 of 2 cohorts to receive 4 treatments: (1) dolutegravir alone, fasting; (2) dolutegravir with calcium carbonate or ferrous fumarate, fasting; (3) dolutegravir with calcium carbonate or ferrous fumarate with a moderate-fat meal; (4) dolutegravir administered 2 hours before calcium carbonate or ferrous fumarate, fasting. Plasma dolutegravir AUC(0-∞), Cmax , and C24 were reduced by 39%, 37%, and 39%, respectively, when co-administered with calcium carbonate while fasting and were reduced by 54%, 57%, and 56%, respectively, when co-administered with ferrous fumarate while fasting. Dolutegravir administration 2 hours before calcium or iron supplement administration (fasted), as well as administration with a meal, counteracted the effect. Dolutegravir and calcium or iron supplements can be co-administered if taken with a meal. Under fasted conditions, dolutegravir should be administered 2 hours before or 6 hours after calcium or iron supplements.
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis