Merck
  • Expression of inactive glutathione peroxidase 4 leads to embryonic lethality, and inactivation of the Alox15 gene does not rescue such knock-in mice.
Expression of inactive glutathione peroxidase 4 leads to embryonic lethality, and inactivation of the Alox15 gene does not rescue such knock-in mice.
Antioxidants & redox signaling (2014-10-15)
Simone Hanna Brütsch, Chi Chiu Wang, Lu Li, Hannelore Stender, Nilgün Neziroglu, Constanze Richter, Hartmut Kuhn, Astrid Borchert
ABSTRACT
Glutathione peroxidases (Gpx) and lipoxygenases (Alox) are functional counterplayers in the metabolism of hydroperoxy lipids that regulate cellular redox homeostasis. Gpx4 is a moonlighting protein that has been implicated not only as an enzyme in anti-oxidative defense, gene expression regulation, and programmed cell death, but also as a structural protein in spermatogenesis. Homozygous Gpx4 knock-out mice are not viable, but molecular reasons for intrauterine lethality are not completely understood. This study was aimed at investigating whether the lack of catalytic activity or the impaired function as structural protein is the dominant reason for embryonic lethality. We further explored whether the pro-oxidative enzyme mouse 12/15 lipoxygenase (Alox15) plays a major role in embryonic lethality of Gpx4-deficient mice. To achieve these goals, we first created knock-in mice, which express a catalytically inactive Gpx4 mutant (Sec46Ala). As homozygous Gpx4-knock-out mice Sec46Ala-Gpx4(+/+) knock-in animals are not viable but undergo intrauterine resorption between embryonic day 6 and 7 (E6-7). In contrast, heterozygous knock-in mice (Sec46Ala-Gpx4(-/+)) are viable, fertile and do not show major phenotypic alterations. Interestingly, homozygous Alox15 deficiency did not rescue the U46A-Gpx4(+/+) mice from embryonic lethality. In fact, when heterozygous U46A-Gpx4(-/+) mice were stepwise crossed into an Alox15-deficent background, no viable U46A-Gpx4(+/+)+Alox15(-/-) individuals were obtained. However, we were able to identify U46A-Gpx4(+/+)+Alox15(-/-) embryos in the state of resorption around E7. These data suggest that the lack of catalytic activity is the major reason for the embryonic lethality of Gpx4(-/-) mice and that systemic inactivation of the Alox15 gene does not rescue homozygous knock-in mice expressing catalytically silent Gpx4.
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Selenium, pellets, < 5mm, ≥99.999%
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane