Merck
  • Detection and characterization of triamcinolone acetonide metabolites in human urine by liquid chromatography/tandem mass spectrometry after intramuscular administration.
Detection and characterization of triamcinolone acetonide metabolites in human urine by liquid chromatography/tandem mass spectrometry after intramuscular administration.
Rapid communications in mass spectrometry : RCM (2015-01-07)
Xavier Matabosch, Oscar J Pozo, Esther Papaseit, Magí Farré, Josep Marcos, Jordi Segura, Rosa Ventura
ABSTRACT
Glucocorticosteroids are prohibited in sports when used by systemic administrations (e.g. intramuscular, IM), whereas they are allowed using other ways of administration. Strategies to discriminate between administrations routes have to be developed by doping control laboratories. For this reason, the metabolism of triamcinolone acetonide (TA), one of the most used glucocorticosteroids, was studied using liquid chromatography coupled to tandem mass spectrometry (LC/MS/MS). Urine samples obtained after IM administration of TA were analyzed using two sample treatments: (a) hydrolysis with β-glucuronidase enzymes and liquid-liquid extraction under alkaline conditions, and (b) liquid-liquid extraction under acidic conditions. The extracts were analyzed by LC/MS/MS. TA, commercially available metabolites (6β-hydroxytriamcinolone acetonide, 6β-OH-TA, and triamcinolone), and their C20-reduced derivatives showed characteristic fragmentation behavior. Besides common product ions and neutral losses for corticosteroids containing fluorine, additional characteristic neutral losses (58 Da, loss of acetone; 44 Da, loss of acetaldehyde) were observed in positive electrospray ionization. Based on that behavior, two complementary approaches were applied to detect TA metabolites: (a) open detection by precursor ion and neutral loss scan methods and (b) targeted detection by selected reaction monitoring methods (SRM) containing theoretical ion transitions of the potential metabolites. Two main compounds, TA and 6β-OH-TA, and nine minor potential metabolites, were detected by open screening methods. Using SRM, two additional metabolites were detected. Some of the metabolites were characterized using reference standards and, for the rest of metabolites, feasible structures were proposed based on mass spectrometric data. Metabolites resulting from hydroxylation in C-6, oxidation of the 11-hydroxyl group, reduction of the Δ(4) double bond and oxidation of the side chain were detected. Some of them have not been previously described. Excretion profiles of the detected metabolites after IM administration are presented.
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C