Merck
  • Cell-penetrating apoptotic peptide/p53 DNA nanocomplex as adjuvant therapy for drug-resistant breast cancer.
Cell-penetrating apoptotic peptide/p53 DNA nanocomplex as adjuvant therapy for drug-resistant breast cancer.
Molecular pharmaceutics (2014-08-01)
Huiyuan Wang, Huixin Wang, Jianming Liang, Yifan Jiang, Qianqian Guo, Huige Peng, Qin Xu, Yongzhuo Huang
ABSTRACT
Drug resistance becomes a formidable challenge against effective cancer therapy. Defective apoptosis in cancer cells is a key factor responsible for chemoresistance or radioresistance. Promoting apoptosis is an important method to sensitize the resistant cells, thereby achieving successful treatment for MDR cancer. We present a strategy of codelivery of apoptotic AVPI peptide and p53 DNA as apoptosis-induction adjuvant therapy for combating the resistant breast cancer. AVPI tetrapeptide is poorly cell-permeable, thereby with very limited value for therapeutic use. Cell-penetrating chimeric AVPI derivative was developed by modification with an octa-arginine sequence (R8). The AVPIR8 is able to not only efficiently penetrate into tumor cells but also work as a vector for gene delivery by forming nanocomplexes based on its cationic R8 moiety. The combination of AVPIR8/p53 DNA was selected for targeting apoptotic pathways, thereby sensitizing the cancer cells to chemotherapeutics. The anti-MDR effect was demonstrated both in vitro and in vivo. The synergistic use of AVPIR8/p53 significantly increased the sensitivity of the resistant tumor cells to the cytotoxic agent doxorubicin by inducing apoptosis, as demonstrated in the cellular studies. Importantly, the treatment improvement was also observed in the animal studies with resistant breast tumor model. Coadministration of AVPIR8/p53 enabled a full arrest of tumor growth combined with a reduced DOX dose, yielding a productive and safe cancer treatment.
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Fmoc-Pro-OH, ≥90% (HPLC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Fmoc-Ala-OH, 95%
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Triisopropylsilane, 98%
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Fmoc-Val-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
N,N-Diisopropylethylamine, 99.5%, biotech. grade