Merck
  • Methanolic extract of Origanum vulgare ameliorates type 1 diabetes through antioxidant, anti-inflammatory and anti-apoptotic activity.
Methanolic extract of Origanum vulgare ameliorates type 1 diabetes through antioxidant, anti-inflammatory and anti-apoptotic activity.
The British journal of nutrition (2015-02-12)
Milica Vujicic, Ivana Nikolic, Vassiliki G Kontogianni, Tamara Saksida, Pantelis Charisiadis, Zorana Orescanin-Dusic, Dusko Blagojevic, Stanislava Stosic-Grujicic, Andreas G Tzakos, Ivana Stojanovic
ABSTRACT
Type 1 diabetes (T1D), an autoimmune inflammatory disorder, develops as a consequence of pancreatic β-cell destruction and results in hyperglycaemia. Since current T1D therapy mainly involves insulin replacement, the aim of the present study was to evaluate the therapeutic potential of Origanum vulgare L. ssp. hirtum (Greek oregano) leaf extract rich in biophenols for the treatment of T1D. The phytochemical profile of methanolic oregano extract (MOE) and aqueous oregano extract (AOE) was determined by liquid chromatography/electrospray ion-trap tandem MS (LC/DAD/ESI-MSn), while their main compounds were quantified by HPLC with diode array detection. After establishing their potent in vitro antioxidant activity, the extracts were administered to C57BL/6 mice treated with multiple low doses of streptozotocin for diabetes induction. While prophylactic AOE therapy had no impact on diabetes induction, MOE reduced diabetes incidence and preserved normal insulin secretion. In addition, MOE scavenged reactive oxygen and nitrogen species and, therefore, alleviated the need for the up-regulation of antioxidant enzymes. MOE treatment specifically attenuated the pro-inflammatory response mediated by T helper 17 cells and enhanced anti-inflammatory T helper 2 and T regulatory cells through the impact on specific signalling pathways and transcription factors. Importantly, MOE preserved β-cells from in vitro apoptosis via blockade of caspase 3. Finally, rosmarinic acid, a predominant compound in MOE, exhibited only partial protection from diabetes induction. In conclusion, acting as an antioxidant, immunomodulator and in an anti-apoptotic manner, MOE protected mice from diabetes development. Seemingly, there is more than one compound responsible for the beneficial effect of MOE.
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Rosmarinic acid, primary reference standard
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Naringenin, natural (US), 98%
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
(±)-Naringenin, analytical standard
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
(±)-Naringenin, ≥95%
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Rosmarinic acid, 96%
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether