Merck
  • Interaction between p53 mutation and a somatic HDMX biomarker better defines metastatic potential in breast cancer.
Interaction between p53 mutation and a somatic HDMX biomarker better defines metastatic potential in breast cancer.
Cancer research (2015-02-05)
Anna M Grawenda, Elen K Møller, Suzanne Lam, Emmanouela Repapi, Amina F A S Teunisse, Grethe I G Alnæs, Anne-Lise Børresen-Dale, Vessela N Kristensen, Colin R Goding, Aart G Jochemsen, Hege Edvardsen, Gareth L Bond
ABSTRACT
TP53 gene mutation is associated with poor prognosis in breast cancer, but additional biomarkers that can further refine the impact of the p53 pathway are needed to achieve clinical utility. In this study, we evaluated a role for the HDMX-S/FL ratio as one such biomarker, based on its association with other suppressor mutations that confer worse prognosis in sarcomas, another type of cancer that is surveilled by p53. We found that HDMX-S/FL ratio interacted with p53 mutational status to significantly improve prognostic capability in patients with breast cancer. This biomarker pair offered prognostic utility that was comparable with a microarray-based prognostic assay. Unexpectedly, the utility tracked independently of DNA-damaging treatments and instead with different tumor metastasis potential. Finally, we obtained evidence that this biomarker pair might identify patients who could benefit from anti-HDM2 strategies to impede metastatic progression. Taken together, our work offers a p53 pathway marker, which both refines our understanding of the impact of p53 activity on prognosis and harbors potential utility as a clinical tool.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nutlin-3, ≥98% (HPLC), powder
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard