Merck
  • NBD-based fluorescent chemosensor for the selective quantification of copper and sulfide in an aqueous solution and living cells.
NBD-based fluorescent chemosensor for the selective quantification of copper and sulfide in an aqueous solution and living cells.
Organic & biomolecular chemistry (2015-01-09)
Qingtao Meng, Yu Shi, Cuiping Wang, Hongmin Jia, Xue Gao, Run Zhang, Yongfei Wang, Zhiqiang Zhang
ABSTRACT
Chemosensors play important roles in cation and anion recognition in biological, industrial, and environmental processes. Although many efforts have been made to develop artificial fluorescent receptors for Cu(2+) and S(2-), their applications in the detection in bulk solutions are limited. In this work, we report a novel fluorescence chemosensor (NL) based on the 7-nitrobenz-2-oxa-1,3-diazole (NBD) fluorophore for the quantification of Cu(2+) and S(2-) in single intact cells. NL specifically binds to Cu(2+) in the presence of other competing cations, and evident changes in UV-vis and fluorescence spectra in HEPES buffer are noticed. Based on the displacement approach, the selective sense S(2-) with the in situ generated NL-Cu(2+) ensemble gives a remarkable recovery of fluorescence and UV-vis absorption spectra. The detection limits of NL for Cu(2+) and NL-Cu(2+) for S(2-) were estimated to be 1.6 nM and 0.17 μM, respectively. NL and the resultant complex NL-Cu(2+) exhibit low cytotoxicity and cell-membrane permeability, which makes them capable of Cu(2+) and S(2-) imaging and quantification in living MDA-MB-231 cells.
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
4-(Diethylamino)salicylaldehyde, 98%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture