Merck
  • Oligodendrocyte precursor cells support blood-brain barrier integrity via TGF-β signaling.
Oligodendrocyte precursor cells support blood-brain barrier integrity via TGF-β signaling.
PloS one (2014-08-01)
Ji Hae Seo, Takakuni Maki, Mitsuyo Maeda, Nobukazu Miyamoto, Anna C Liang, Kazuhide Hayakawa, Loc-Duyen D Pham, Fumihiko Suwa, Akihiko Taguchi, Tomohiro Matsuyama, Masafumi Ihara, Kyu-Won Kim, Eng H Lo, Ken Arai
ABSTRACT
Trophic coupling between cerebral endothelium and their neighboring cells is required for the development and maintenance of blood-brain barrier (BBB) function. Here we report that oligodendrocyte precursor cells (OPCs) secrete soluble factor TGF-β1 to support BBB integrity. Firstly, we prepared conditioned media from OPC cultures and added them to cerebral endothelial cultures. Our pharmacological experiments showed that OPC-conditioned media increased expressions of tight-junction proteins and decreased in vitro BBB permeability by activating TGB-β-receptor-MEK/ERK signaling pathway. Secondly, our immuno-electron microscopic observation revealed that in neonatal mouse brains, OPCs attach to cerebral endothelial cells via basal lamina. And finally, we developed a novel transgenic mouse line that TGF-β1 is knocked down specifically in OPCs. Neonates of these OPC-specific TGF-β1 deficient mice (OPC-specific TGF-β1 partial KO mice: PdgfraCre/Tgfb1flox/wt mice or OPC-specific TGF-β1 total KO mice: PdgfraCre/Tgfb1flox/flox mice) exhibited cerebral hemorrhage and loss of BBB function. Taken together, our current study demonstrates that OPCs increase BBB tightness by upregulating tight junction proteins via TGF-β signaling. Although astrocytes and pericytes are well-known regulators of BBB maturation and maintenance, these findings indicate that OPCs also play a pivotal role in promoting BBB integrity.
Sigma-Aldrich
DAPI, for nucleic acid staining