Merck
Effect of sterilization and crosslinking on gelatin films.
Journal of materials science. Materials in medicine (2015-01-30)
Sofia Amadori, Paola Torricelli, Katia Rubini, Milena Fini, Silvia Panzavolta, Adriana Bigi
ABSTRACT
Sterilization through γ-irradiation has been reported to affect collagen mechanical properties, but its possible effects on gelatin based materials have not been investigated up to now. Herein we report the results of a mechanical, chemical and thermal study performed on gelatin films before and after γ-irradiation. The investigation was performed on uncrosslinked films as well as on crosslinked films. To this aim, two common crosslinking agents, glutaraldehyde and genipin, at different concentration (0.15, 0.30 and 0.67%) were used. The results indicate that sterilization significantly affects the mechanical properties of uncrosslinked films, whereas it displays a modest effect on gelatin swelling, release in solution, thermal stability and molecular structure. Both glutaraldehyde and genipin enhance the mechanical properties and stability in solution of the gelatin films. In particular, the values of Young modulus increase as a function of crosslinker concentration up to about 10 and 18 MPa for genipin and glutaraldehyde treated samples respectively. The results of in vitro study demonstrate that the films crosslinked with genipin do not display any cytotoxic reaction, whereas glutaraldehyde crosslinking provokes an acute and dose dependent cytotoxic effect.
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Decane, anhydrous, ≥99%
Sigma-Aldrich
Decane, ≥95%
Supelco
Decane, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Copper(II) sulfate pentahydrate, purum p.a., crystallized, ≥99.0% (RT)
Supelco
Decane, analytical standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Decane, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Copper(II) sulfate pentahydrate, puriss., suitable for, meets analytical specification of Ph. Eur., BP, USP,FCC
Sigma-Aldrich
Copper(II) sulfate pentahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99-102%