Merck
  • Organic cation transporter/solute carrier family 22a is involved in drug transfer into milk in mice.
Organic cation transporter/solute carrier family 22a is involved in drug transfer into milk in mice.
Journal of pharmaceutical sciences (2014-09-02)
Naoki Ito, Kousei Ito, Yuki Ikebuchi, Tomoko Kito, Hiroshi Miyata, Yu Toyoda, Tappei Takada, Akihiro Hisaka, Masashi Honma, Akira Oka, Hiroyuki Kusuhara, Hiroshi Suzuki
ABSTRACT
Drug transfer into milk is a general concern during lactation. So far, breast cancer resistance protein (Bcrp) is the only transporter known to be involved in this process, whereas participation of other transporters remains unclear. We investigated the importance of organic cation transporter (Oct) in drug transfer into milk in mice. The mammary glands of lactating versus nonlactating FVB strain mice revealed elevated mRNA levels of Oct1 and Bcrp, whereas Oct2 and Oct3 mRNA levels were decreased. Specific uptake of cimetidine, acyclovir, metformin, and terbutaline was observed in human embryonic kidney 293 cells transfected with murine Oct1 or Oct2. The milk-to-plasma concentration ratio (M/P) values of cimetidine and acyclovir were significantly decreased in Bcrp knockout and Oct1/2 double-knockout (DKO) mice compared with control FVB mice, whereas the M/P values of terbutaline and metformin were significantly decreased in Oct1/2 DKO mice alone. These are the first to suggest that Oct1 might be involved in secretory transfer of substrate drugs into milk.
Sigma-Aldrich
Acycloguanosine, ≥99% (HPLC), powder
Sigma-Aldrich
Terbutaline hemisulfate salt
Sigma-Aldrich
Cimetidine
Aciclovir, European Pharmacopoeia (EP) Reference Standard
Supelco
Cimetidine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acyclovir, Pharmaceutical Secondary Standard; Certified Reference Material
Cimetidine, European Pharmacopoeia (EP) Reference Standard
Clindamycin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Terbutaline sulfate, European Pharmacopoeia (EP) Reference Standard
Cimetidine for peak identification, European Pharmacopoeia (EP) Reference Standard
Cimetidine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Clindamycin hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Clindamycin hydrochloride, lincosamide antibiotic