Merck
Long non-coding RNA deregulation in tongue squamous cell carcinoma.
BioMed research international (2014-07-22)
Wei Gao, Jimmy Yu-Wai Chan, Thian-Sze Wong
ABSTRACT
The deregulated tumorigenic long non-coding RNA (lncRNA) has been reported in several malignancies. However, there is still no comprehensive study on tongue squamous cell carcinoma (SCC). Functional reannotation for the human lncRNA was carried out by ncFANs. Real-time quantitative PCR was used to validate the identified lncRNAs. Using the functional annotation algorithm from ncFANs, 8 differentially expressed lncRNAs were identified. Lnc-PPP2R4-5, lnc-SPRR2D-1, lnc-MAN1A2-1, lnc-FAM46A-1, lnc-MBL2-4:1, and lnc-MBL2-4:3 were upregulated in the microdissected tongue SCC tissues. In comparison, lnc-AL355149.1-1 and lnc-STXBP5-1 showed significant downregulation. High level of lnc-MBL2-4:3 was significantly associated with the node positive tongue SCC patients. Further, patients with advanced T-stage demonstrated a further reduction of lnc-AL355149.1-1 in the tumor tissues. Treatment of tongue SCC cells with 5-fluorouracil and paclitaxel can reserve the expression patterns observed in the tongue SCC tissues. Further, changes of lnc-MBL2-4:3 and lnc-AL355149.1-1 expression levels were noticed in the cisplatin-resistant tongue SCC cells. Our results demonstrated that functional reannotation allows us to identify novel lncRNAs using the existing gene expression array dataset. The association of lncRNA with the T-stage and nodal status of tongue SCC patients suggested that lncRNA deregulation was involved in the pathogenesis of tongue SCC.
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
5-Fluorouracil, analytical standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water