Merck
  • Two-stage chromatographic separation of aggregates for monoclonal antibody therapeutics.
Two-stage chromatographic separation of aggregates for monoclonal antibody therapeutics.
Journal of chromatography. A (2014-12-03)
Vijesh Kumar, Anurag S Rathore
ABSTRACT
Aggregates of monoclonal antibody (mAb) therapeutics, due to their perceived impact on immunogenicity, are recognized as a critical quality attribute by the regulatory authorities as well as the industry. Hence, removal of aggregates is a key objective of bioprocessing. At present, this is achieved by a combination of two or more orthogonal chromatographic steps with possible modalities of ion exchange, hydrophobic interaction and mixed mode. A two-stage chromatographic purification process consisting of ion-exchange and hydrophobic interaction modes is proposed in this paper for effective and efficient control of aggregates for a mAb therapeutic. The proposed scheme does not require any intermediate processing of the process stream. Further, baseline separation is achieved for monomer and aggregates resulting in robust performance. This was possible because the proposed operational scheme allowed for an addition of selectivities of the two chromatography modes vs. the traditional two column scheme where part of the separation of aggregates achieved by the first column is lost upon pooling. The proposed process scheme yielded improved separation of aggregates (0% vs. 1-2%) at >95% recovery and reduced overall process time (6h vs. 14 h) for a typical application. Further, clearance of host cell proteins was also shown to have improved with the suggested process scheme. Successful implementation of the proposed scheme has been demonstrated for two different monoclonal antibody therapeutic products.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
SAFC
HEPES
SAFC
HEPES
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%