Merck
  • Silica nanoparticles as the adjuvant for the immunisation of mice using hepatitis B core virus-like particles.
Silica nanoparticles as the adjuvant for the immunisation of mice using hepatitis B core virus-like particles.
PloS one (2014-12-02)
Dace Skrastina, Ivars Petrovskis, Ilva Lieknina, Janis Bogans, Regina Renhofa, Velta Ose, Andris Dishlers, Yuri Dekhtyar, Paul Pumpens
ABSTRACT
Advances in nanotechnology and nanomaterials have facilitated the development of silicon dioxide, or Silica, particles as a promising immunological adjuvant for the generation of novel prophylactic and therapeutic vaccines. In the present study, we have compared the adjuvanting potential of commercially available Silica nanoparticles (initial particles size of 10-20 nm) with that of aluminium hydroxide, or Alum, as well as that of complete and incomplete Freund's adjuvants for the immunisation of BALB/c mice with virus-like particles (VLPs) formed by recombinant full-length Hepatitis B virus core (HBc) protein. The induction of B-cell and T-cell responses was studied after immunisation. Silica nanoparticles were able to adsorb maximally 40% of the added HBc, whereas the adsorption capacity of Alum exceeded 90% at the same VLPs/adjuvant ratio. Both Silica and Alum formed large complexes with HBc VLPs that sedimented rapidly after formulation, as detected by dynamic light scattering, spectrophotometry, and electron microscopy. Both Silica and Alum augmented the humoral response against HBc VLPs to the high anti-HBc level in the case of intraperitoneal immunisation, whereas in subcutaneous immunisation, the Silica-adjuvanted anti-HBc level even exceeded the level adjuvanted by Alum. The adjuvanting of HBc VLPs by Silica resulted in the same typical IgG2a/IgG1 ratios as in the case of the adjuvanting by Alum. The combination of Silica with monophosphoryl lipid A (MPL) led to the same enhancement of the HBc-specific T-cell induction as in the case of the Alum and MPL combination. These findings demonstrate that Silica is not a weaker putative adjuvant than Alum for induction of B-cell and T-cell responses against recombinant HBc VLPs. This finding may have an essential impact on the development of the set of Silica-adjuvanted vaccines based on a long list of HBc-derived virus-like particles as the biological component.
Product Description
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Mouse Monoclonal Antibody Isotyping Reagents, sufficient for 1000 tests (clones) (by ELISA), sufficient for 40 tests (clones) (by immunodiffusion, ODD)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Pricing and availability is not currently available. Contact Technical Service
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Urea, meets USP testing specifications
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Urea, suitable for electrophoresis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Pricing and availability is not currently available. Contact Technical Service
Urea, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available. Contact Technical Service