Merck
  • Derivation of keratinocytes from chicken embryonic stem cells: establishment and characterization of differentiated proliferative cell populations.
Derivation of keratinocytes from chicken embryonic stem cells: establishment and characterization of differentiated proliferative cell populations.
Stem cell research (2015-02-24)
Mathilde Couteaudier, Laëtitia Trapp-Fragnet, Nicolas Auger, Katia Courvoisier, Bertrand Pain, Caroline Denesvre, Jean-François Vautherot
ABSTRACT
A common challenge in avian cell biology is the generation of differentiated cell-lines, especially in the keratinocyte lineage. Only a few avian cell-lines are available and very few of them show an interesting differentiation profile. During the last decade, mammalian embryonic stem cell-lines were shown to differentiate into almost all lineages, including keratinocytes. Although chicken embryonic stem cells had been obtained in the 1990s, few differentiation studies toward the ectodermal lineage were reported. Consequently, we explored the differentiation of chicken embryonic stem cells toward the keratinocyte lineage by using a combination of stromal induction, ascorbic acid, BMP4 and chicken serum. During the induction period, we observed a downregulation of pluripotency markers and an upregulation of epidermal markers. Three homogenous cell populations were derived, which were morphologically similar to chicken primary keratinocytes, displaying intracellular lipid droplets in almost every pavimentous cell. These cells could be serially passaged without alteration of their morphology and showed gene and protein expression profiles of epidermal markers similar to chicken primary keratinocytes. These cells represent an alternative to the isolation of chicken primary keratinocytes, being less cumbersome to handle and reducing the number of experimental animals used for the preparation of primary cells.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Pricing and availability is not currently available.
Supelco
L-Ascorbic acid, analytical standard
Pricing and availability is not currently available.
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Pricing and availability is not currently available.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Pricing and availability is not currently available.
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Pricing and availability is not currently available.
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Adenine, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Adenine hydrochloride hydrate, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Adenine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Selenium, pellets, < 5mm, ≥99.999%
Pricing and availability is not currently available.