Merck
  • Supported inhibitor for fishing lipases in complex biological media and mass spectrometry identification.
Supported inhibitor for fishing lipases in complex biological media and mass spectrometry identification.
Biochimie (2014-07-30)
Vincent Delorme, Brigitt Raux, Rémy Puppo, Julien Leclaire, Jean-François Cavalier, Sylvain Marc, Pavan-Kumar Kamarajugadda, Gérard Buono, Frédéric Fotiadu, Stéphane Canaan, Frédéric Carrière
ABSTRACT
A synthetic phosphonate inhibitor designed for lipase inhibition but displaying a broader range of activity was covalently immobilized on a solid support to generate a function-directed tool targeting serine hydrolases. To achieve this goal, straightforward and reliable analytical techniques were developed, allowing the monitoring of the solid support's chemical functionalization, enzyme capture processes and physisorption artifacts. This grafted inhibitor was tested on pure lipases and serine proteases from various origins, and assayed for the selective capture of lipases from several complex biological extracts. The direct identification of captured enzymes by mass spectrometry brought the proof of concept on the efficiency of this supported covalent inhibitor. The features and limitations of this "enzyme-fishing" proteomic tool provide new insight on solid-liquid inhibition process.
Sigma-Aldrich
3-Indoleacetic acid, 98%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Nα-Benzoyl-L-arginine ethyl ester hydrochloride, trypsin substrate
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Supelco
3-Indoleacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%