Merck
  • Liquid chromatography-mass spectrometry analysis of diethylcarbamazine in human plasma for clinical pharmacokinetic studies.
Liquid chromatography-mass spectrometry analysis of diethylcarbamazine in human plasma for clinical pharmacokinetic studies.
Journal of pharmaceutical and biomedical analysis (2014-07-01)
Mark S Schmidt, Christopher L King, Edward K Thomsen, Peter M Siba, Nelly Sanuku, Lawrence Fleckenstein
ABSTRACT
A sensitive and selective liquid chromatographic method using mass spectrometric detection was developed for the determination of diethylcarbamazine (DEC) in human plasma. DEC and its stable isotope internal standard d3-DEC were extracted from 0.25mL of human plasma using solid phase extraction. Chromatography was performed using a Phenomenex Synergi 4μ Fusion-RP column (2mm×250mm) with gradient elution. The retention time was approximately 4.8min. The assay was linear from 4 to 2200ng/mL. Analysis of quality control samples at 12, 300, and 1700ng/mL (N=15) had interday coefficients of variation of 8.4%, 5.4%, and 6.2%, respectively (N=15). Interday bias results were -2.2%, 6.0%, and 0.8%, respectively. Recovery of DEC from plasma ranged from 84.2% to 90.1%. The method was successfully applied to clinical samples from patients with lymphatic filariasis from a drug-drug interaction study between DEC and albendazole and/or ivermectin.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Diethylcarbamazine citrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material