Merck
  • Differential stimulation of peripheral blood mononuclear cells in Crohn's disease by fungal glycans.
Differential stimulation of peripheral blood mononuclear cells in Crohn's disease by fungal glycans.
Journal of gastroenterology and hepatology (2014-08-06)
Liran Baram, Sarit Cohen-Kedar, Lior Spektor, Hofit Elad, Hanan Guzner-Gur, Iris Dotan
ABSTRACT
Crohn's disease (CD) is characterized by loss of tolerance to intestinal microorganisms. This is reflected by serological responses to fungal glycans such as mannan and β-glucans. Fungal glycans have various effects on immune cells. However, the evidence for their effects in CD is vague. This study aimed to assess the effects of fungal cell wall glycans on human peripheral blood mononuclear cells (PBMCs) from CD and control patients. Human PBMCs from CD and control patients were stimulated by fungal cell wall glycans. Cytokine secretion was detected by ELISA and glycan receptor expression by flow cytometry. Mannan, β-glucans (curdlan), chitosan, and zymosan induced the secretion of interleukin (IL)-1β, IL-6, IL-23, IL-10, and tumor necrosis factor-α by PBMCs. Spleen tyrosin kinase and Src tyrosine kinase were involved in the response to mannan and β-glucans. Mannan and whole yeast cells induced a significantly higher pro-inflammatory cytokine response in CD compared with control patients. The results may suggest that CD is characterized by hyperresponsiveness to fungal glycans. Thus, glycans may potentially be triggering or perpetuating inflammation.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
SAFC
L-Tyrosine
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Piceatannol, powder
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
L-Tyrosine, FG
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material