Merck
  • Influence of copolymer composition on the phase behavior of solid dispersions.
Influence of copolymer composition on the phase behavior of solid dispersions.
Molecular pharmaceutics (2014-10-09)
Anke Prudic, Tobias Kleetz, Marcel Korf, Yuanhui Ji, Gabriele Sadowski
ABSTRACT
The incorporation of poorly soluble active pharmaceutical ingredients (APIs) into excipients (e.g., polymers) to formulate an amorphous solid dispersion is a promising strategy to improve the oral bioavailability of the API. The application of copolymer excipients allows access to combinations of different monomers and thus to the design of excipients to improve solid-dispersion properties. In this work, the thermodynamic phase behavior of solid dispersions was investigated as a function of the API, type of monomer, and copolymer composition. The glass-transition temperatures and API solubilities in the solid dispersions of naproxen and indomethacin in polyvinylpyrrolidone, polyvinyl acetate, and copolymers with different weight fractions of vinylpyrrolidone and vinyl actetate were investigated. It is shown that the thermodynamic phase behavior of API/copolymer solid dispersions is a function of monomer type and copolymer composition. This effect was also predicted by using the perturbed-chain statistical associating fluid theory (PC-SAFT). The glass-transition temperature of the solid dispersions was calculated with the Gordon-Taylor equation.
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Naproxen, VETRANAL®, analytical standard
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Sigma-Aldrich
Naproxen, meets USP testing specifications
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Naproxen, European Pharmacopoeia (EP) Reference Standard