Merck
  • Identification of human cytochrome P450 isozymes involved in the metabolism of naftopidil enantiomers in vitro.
Identification of human cytochrome P450 isozymes involved in the metabolism of naftopidil enantiomers in vitro.
The Journal of pharmacy and pharmacology (2014-06-20)
Lijun Zhu, Xiawen Liu, Liu Zhu, Xingfei Zhang, Xiaojing Fu, Junjun Huang, Mu Yuan
ABSTRACT
Naftopidil (NAF) is a chiral compound with two enantiomers (R(+)-NAF and S(-)-NAF) and is used as a racemic mixture in clinical practice. This study aims to investigate the metabolism of NAF enantiomers in pooled human liver microsomes (HLMs) and cytochrome P450 isozymes (CYPs) involved in their metabolism. Metabolism studies were conducted in vitro using HLMs. Specific chemical inhibitors and recombinant human CYPs were used to confirm that the CYPs contributed to the metabolism of NAF enantiomers. Three metabolites were found and characterized in the HLMs incubations from R(+)-NAF and S(-)-NAF, respectively. The major metabolic pathways of R(+)-NAF and S(-)-NAF were demethylation and hydroxylation. CYP2C9 and CYP2C19 inhibitors strongly inhibited R(+)-NAF metabolism, and CYP1A2, CYP2C8, CYP2D6 and CYP3A4/5 inhibitors moderately inhibited R(+)-NAF metabolism. CYP2C9 inhibitors strongly inhibited S(-)-NAF metabolism, and CYP2C8, CYP2C19 and CYP3A4/5 inhibitors moderately inhibited S(-)-NAF metabolism. Consistent with the results of chemical inhibitors experiments, recombinant human CYP2C9 and CYP2C19 contributed greatly to R(+)-NAF metabolism, and CYP2C9 contributed greatly to S(-)-NAF metabolism. Both R(+)-NAF and S(-)-NAF are metabolized to three metabolites in HLMs. CYP2C9 plays the most important role in the demethylation and hydroxylation of both NAF enantiomers, CYP2C19 is another major CYP isoform that is involved in R(+)-NAF metabolism.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Supelco
Ticlopidine hydrochloride, analytical standard, for drug analysis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Ticlopidine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%