Merck
  • The role of microRNA-15b in the impaired angiogenesis in diabetic wounds.
The role of microRNA-15b in the impaired angiogenesis in diabetic wounds.
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society (2014-07-26)
Junwang Xu, Carlos Zgheib, Junyi Hu, Wenjie Wu, Liping Zhang, Kenneth W Liechty
ABSTRACT
The impairment in diabetic wound healing represents a significant clinical problem. Decreased angiogenesis is thought to play a central role in the pathogenesis of this impairment. We have previously shown that treatment of diabetic murine wounds with mesenchymal stem cells can improve healing, but the mechanisms are not completely defined. MicroRNA-15b (miR-15b) has been implicated in the regulation of the angiogenic response. We hypothesized that abnormal miR-15b expression may contribute to the impaired angiogenesis observed in impaired diabetic wound healing. To test this hypothesis, we examined the expression of miR-15b and its target genes in diabetic and nondiabetic mice before and after injury. MiR-15b expression was significantly up-regulated in diabetic mouse wounds during the wound healing response. Increased miR-15b levels also closely correlated with decreased gene expression of its proangiogenic target genes. Furthermore, the correction of the diabetic wound healing impairment with mesenchymal stem cell treatment was associated with a significant decrease in miR-15b expression level and increased gene expression of its proangiogenic target genes. These results provide the first evidence that increased expression of miR-15b in diabetic wounds in response to injury may, in part, be responsible for the abnormal angiogenic response seen in diabetic wounds and may contribute to the observed wound healing impairment.
Sigma-Aldrich
Flunixin meglumine, ≥98% (HPLC)
Supelco
Flunixin Meglumine, Pharmaceutical Secondary Standard; Certified Reference Material