Merck
  • Identification of a neuroprotective and selective butyrylcholinesterase inhibitor derived from the natural alkaloid evodiamine.
Identification of a neuroprotective and selective butyrylcholinesterase inhibitor derived from the natural alkaloid evodiamine.
European journal of medicinal chemistry (2014-05-14)
Guozheng Huang, Beata Kling, Fouad H Darras, Jörg Heilmann, Michael Decker
ABSTRACT
Two sets of carbamates based on the natural alkaloid evodiamine were designed, synthesized and evaluated as potential butyrylcholinesterase inhibitors. Although a set of carbamates of 3-hydroxyevodiamine (10a-f) is inactive both at AChE and BChE, carbamates of 5-deoxo-3-hydroxyevodiamine (11a-f) exhibit much better potency with selectivity toward BChE. The heptyl carbamate of 5-deoxo-3-hydroxyevodiamine (11c) shows the best potency with an IC50 value of 77 nM and very good selectivity over AChE. ORAC and cell-based assays indicate 11c owns pronounced antioxidant properties with 1.75 Trolox equivalents and strong neuroprotection even from 1 μM onwards. These combined activities might enable compound 11c to be a potential candidate for treatment of Alzheimer's disease.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Fluorescein, free acid
Fluorescein, European Pharmacopoeia (EP) Reference Standard