Merck
  • A biodistribution study of PEGylated PCL-based nanoparticles in C57BL/6 mice bearing B16/F10 melanoma.
A biodistribution study of PEGylated PCL-based nanoparticles in C57BL/6 mice bearing B16/F10 melanoma.
Nanotechnology (2014-07-31)
M Lupi, C Colombo, R Frapolli, R Ferrari, L Sitia, L Dragoni, E Bello, S A Licandro, F Falcetta, P Ubezio, P Bigini, M Salmona, M D'Incalci, M Morbidelli, D Moscatelli
ABSTRACT
One of the major drawbacks that limits the clinical application of nanoparticles is the lack of preliminary investigations related to their biocompatibility, biodegradability and biodistribution. In this work, biodegradable PEGylated polymer nanoparticles (NPs) have been synthesized by using macromonomers based on poly(ε-caprolaconte) oligomers. More in detail, NPs have been produced by adopting a surfactant-free semibatch emulsion polymerization process using PEG chains as a stabilizing agent. The NPs were also labeled with rhodamine B covalently bound to the NPs to quantitatively study their biodistribution in vivo. NPs were investigated in both in vitro and in vivo preclinical systems to study their biodistribution in mice bearing B16/F10 melanoma, as well as their biocompatibility and biodegradability. The NP concentration was evaluated in different tissues at several times after intravenous injection. The disappearance of the NPs from the plasma was biphasic, with distribution and elimination half-lives of 30 min and 15 h, respectively. NPs were retained in tumors and in filter organs for a long time, were still detectable after 7 d and maintained a steady concentration in the tumor for 120 h. 48 h after injection, 70 ± 15% of the inoculated NPs were excreted in the feces. The favorable tumor uptake, fast excretion and absence of cytotoxicity foster the further development of produced NPs as drug delivery carriers.
Sigma-Aldrich
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Rhodamine B, for fluorescence
Supelco
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
Sigma-Aldrich
2-Hydroxyethyl methacrylate, ≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
2-Hydroxyethyl methacrylate, contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Sigma-Aldrich
2-Hydroxyethyl methacrylate, embedding medium (for microscopy)
Supelco
Rhodamine B, analytical standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DCC, 99%
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
DCC, puriss., ≥99.0% (GC)
Sigma-Aldrich
Pentane, anhydrous, ≥99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)