Merck
  • Low doses of ochratoxin A induce micronucleus formation and delay DNA repair in human lymphocytes.
Low doses of ochratoxin A induce micronucleus formation and delay DNA repair in human lymphocytes.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-12-03)
Cyndia A González-Arias, Alma B Benitez-Trinidad, Monserrat Sordo, Lourdes Robledo-Marenco, Irma M Medina-Díaz, Briscia S Barrón-Vivanco, Sonia Marín, Vicente Sanchis, Antonio J Ramos, Aurora E Rojas-García
ABSTRACT
The contamination of food commodities by fungal toxins has attracted great interest because many of these mycotoxins are responsible for different diseases, including cancer and other chronic illnesses. Ochratoxin A (OTA) is a mycotoxin naturally present in food, and long-term exposure to food contaminated with low levels of OTA has been associated with renal cancer. In the present study, the cytotoxicity, cytostaticity, and genotoxicity of OTA (0.075-15 µM) in human lymphocytes were evaluated. A comet assay, a modified comet assay (DNA repair assay), which uses N-hydroxyurea (NHU) to detect non-repaired lesions produced by OTA, and a cytokinesis-blocked micronucleus assay were used. Treatments with OTA were not cytotoxic, but OTA caused a cytostatic effect in human lymphocytes at a concentration of 15 µM. OTA (0.075-5 µM) produced a slight increase in the percentage of DNA in the comets and a delay in the DNA repair capacity of the lymphocytes. Micronucleus (MN) induction was observed at OTA concentrations of 1.5 and 5 µM. Our results indicate that OTA induces DNA stable damage at low doses that are neither cytotoxic nor cytostatic, and OTA delays the DNA repair kinetics. These findings indicate that OTA affects two pivotal events in the carcinogenesis pathway.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%