Merck
  • Metabolism of skin-absorbed resveratrol into its glucuronized form in mouse skin.
Metabolism of skin-absorbed resveratrol into its glucuronized form in mouse skin.
PloS one (2014-12-17)
Itsuo Murakami, Romanas Chaleckis, Tomáš Pluskal, Ken Ito, Kousuke Hori, Masahiro Ebe, Mitsuhiro Yanagida, Hiroshi Kondoh
ABSTRACT
Resveratrol (RESV) is a plant polyphenol, which is thought to have beneficial metabolic effects in laboratory animals as well as in humans. Following oral administration, RESV is immediately catabolized, resulting in low bioavailability. This study compared RESV metabolites and their tissue distribution after oral uptake and skin absorption. Metabolomic analysis of various mouse tissues revealed that RESV can be absorbed and metabolized through skin. We detected sulfated and glucuronidated RESV metabolites, as well as dihydroresveratrol. These metabolites are thought to have lower pharmacological activity than RESV. Similar quantities of most RESV metabolites were observed 4 h after oral or skin administration, except that glucuronidated RESV metabolites were more abundant in skin after topical RESV application than after oral administration. This result is consistent with our finding of glucuronidated RESV metabolites in cultured skin cells. RESV applied to mouse ears significantly suppressed inflammation in the TPA inflammation model. The skin absorption route could be a complementary, potent way to achieve therapeutic effects with RESV.
Sigma-Aldrich
PIPES, ≥99% (titration)
Sigma-Aldrich
PIPES, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
PIPES, BioXtra, ≥99% (titration)
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
PIPES, anhydrous, free-flowing, Redi-Dri, ≥99%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium carbonate, puriss., meets analytical specification of NF, Ph. Franc., FCC
Sigma-Aldrich
Ammonium carbonate, ACS reagent, ≥30.0% NH3 basis
SAFC
HEPES
Sigma-Aldrich
Ammonium carbonate, 99.999% trace metals basis
Sigma-Aldrich
PIPES, BioXtra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O