Merck
  • Synergistic anticancer effects of Pam3CSK4 and Ara-C on B-cell lymphoma cells.
Synergistic anticancer effects of Pam3CSK4 and Ara-C on B-cell lymphoma cells.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-05-07)
Sae-Kyung Lee, Jyh Y Chwee, Cheryl A P Ma, Nina Le Bert, Caleb W Huang, Stephan Gasser
ABSTRACT
The low immunogenicity of many cancer cells and the immunosuppression by various cancers and anticancer therapies have been an obstacle in the development of efficacious immunotherapies. Our goal was to test whether Toll-like receptor (TLR) agonists and anticancer chemotherapeutic agents synergize in rendering tumor cells more immunogenic. We treated B-cell lymphoma cells with the TLR1/2 agonist Pam3CSK4 and the genotoxic anticancer agent 1-β-D-arabinofuranosylcytosine (Ara-C). The effects on the immunogenicity of tumor cells were measured in transfer experiments and in vitro studies. The treatment of B-cell lymphoma cells with the TLR1/2 agonist Pam3CSK4 enhanced the anticancer effects of the genotoxic agent Ara-C. Mice injected with cotreated tumor cells survived longer than mice challenged with Pam3CSK4 or Ara-C-treated cells. Administration of Pam3CSK4 or Ara-C reduced the tumor load of mice injected with tumor cells. Cotreatment had no effect on the rate of apoptosis or proliferation of Ara-C-treated cells, but upregulated the expression of several immunomodulatory molecules. Consistent with an increased immunogenicity of Pam3CSK4 and Ara-C-treated B-cell lymphoma cells, rejection of cotreated tumor cells required natural killer cells and T cells. We demonstrate that the upregulation of immunomodulatory molecules in response to Pam3CSK4 and Ara-C depended in part on NF-κB. TLR agonists can increase the efficacy of conventional cancer therapies by altering the immunogenicity of B-cell lymphoma cells.
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Cytarabine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Anti-IL7R antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human TP53