Merck
Cancer chemotherapy and cardiac arrhythmias: a review.
Drug safety (2015-01-13)
Juan Tamargo, Ricardo Caballero, Eva Delpón
ABSTRACT
Cardiovascular toxicity is a potential complication of cancer chemotherapy (CC) that increases the morbidity and mortality of cancer patients. Cardiac arrhythmias have been reported as an adverse effect of many chemotherapeutic drugs, including novel targeted therapies. The relationship between chemotherapy and arrhythmias has not been well-established and the proarrhythmogenic mechanisms remain uncertain as they can be the result of a direct electrophysiological effect or of changes in cardiac structure and function, including myocardial ischaemia and heart failure, which create an arrhythmogenic substrate. In this review we summarise available evidence of proarrhythmia induced by CC, discuss the possible mechanisms involved in this adverse effect and emphasise the importance of cardiac monitoring for the early diagnosis, intervention and surveillance of those patients more susceptible to develop proarrhythmia in an attempt to reduce the morbidity and mortality. Oncologists should be fully aware of proarrhythmia and the close collaboration between cardiologists and oncologists would result in a better cardiovascular assessment, risk stratification, cardiac monitoring and treatment during CC and during the follow-up. The final objective is to understand the mechanisms of proarrhythmia and evaluate its real incidence and clinical relevance so as to select the safest and most effective treatment for cancer patients.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Carmustine, ≥98%
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Supelco
5-Fluorouracil, analytical standard
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ifosfamide, ≥98%
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Melphalan, powder
Cyclophosphamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bosutinib, ≥98% (HPLC)
Cisplatin, European Pharmacopoeia (EP) Reference Standard