Merck
  • Impaired heart rate regulation and depression of cardiac chronotropic and dromotropic function in polymicrobial sepsis.
Impaired heart rate regulation and depression of cardiac chronotropic and dromotropic function in polymicrobial sepsis.
Shock (Augusta, Ga.) (2014-10-02)
Donald B Hoover, Tammy R Ozment, Robert Wondergem, Chuanfu Li, David L Williams
ABSTRACT
The scope of cardiac pathophysiology in sepsis has not been fully defined. Accordingly, we evaluated the effects of sepsis on heart rate (HR), HR variability, and conduction parameters in a murine model of sepsis. Electrocardiograms were recorded noninvasively from conscious mice before and after cecal ligation and puncture (CLP) or sham surgery. Responses of isolated atria to tyramine and isoproterenol were quantified to assess the functional state of sympathetic nerves and postjunctional sensitivity to adrenergic stimulation. Cecal ligation and puncture mice had lower HR compared with sham at 16 to 18 h postsurgery (sham, 741 ± 7 beats/min; CLP, 557 ± 31 beats/min; n = 6/group; P < 0.001), and there was significant prolongation of the PR, QRS, and QTc intervals. Slowing of HR and conduction developed within 4 to 6 h after CLP and were preceded by a decrease in HR variability. Treatment of CLP mice with isoproterenol (5 mg/kg, intraperitoneally) at 25 h after surgery failed to increase HR or decrease conduction intervals. The lack of in vivo response to isoproterenol cannot be attributed to hypothermia because robust chronotropic and inotropic responses to isoproterenol were evoked from isolated atria at 25 °C and 30 °C. These findings demonstrate that impaired regulation of HR (i.e., reduced HR variability) develops before the onset of overt cardiac rate and conduction changes in septic mice. Subsequent time-dependent decreases in HR and cardiac conduction can be attributed to hypothermia and would contribute to decreased cardiac output and organ perfusion. Because isolated atria from septic mice showed normal responsiveness to adrenergic stimulation, we conclude that impaired effectiveness of isoproterenol in vivo can be attributed to reversible effects of systemic factors on adrenergic receptors and/or postreceptor signaling.
Product Description
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Tyramine hydrochloride, ≥98%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane