Merck
  • Automated image analysis of a glomerular injury marker desmin in spontaneously diabetic Torii rats treated with losartan.
Automated image analysis of a glomerular injury marker desmin in spontaneously diabetic Torii rats treated with losartan.
The Journal of endocrinology (2014-05-02)
Tetsuhiro Kakimoto, Kinya Okada, Yoshihiro Hirohashi, Raissa Relator, Mizue Kawai, Taku Iguchi, Keisuke Fujitaka, Masashi Nishio, Tsuyoshi Kato, Atsushi Fukunari, Hiroyuki Utsumi
ABSTRACT
Diabetic nephropathy is a major complication in diabetes and a leading cause of end-stage renal failure. Glomerular podocytes are functionally and structurally injured early in diabetic nephropathy. A non-obese type 2 diabetes model, the spontaneously diabetic Torii (SDT) rat, is of increasing preclinical interest because of its pathophysiological similarities to human type 2 diabetic complications including diabetic nephropathy. However, podocyte injury in SDT rat glomeruli and the effect of angiotensin II receptor blocker treatment in the early stage have not been reported in detail. Therefore, we have evaluated early stages of glomerular podocyte damage and the beneficial effect of early treatment with losartan in SDT rats using desmin as a sensitive podocyte injury marker. Moreover, we have developed an automated, computational glomerulus recognition method and illustrated its specific application for quantitatively studying glomerular desmin immunoreactivity. This state-of-the-art method enabled automatic recognition and quantification of glomerular desmin-positive areas, eliminating the need to laboriously trace glomerulus borders by hand. The image analysis method not only enabled assessment of a large number of glomeruli, but also clearly demonstrated that glomerular injury was more severe in the juxtamedullary region than in the superficial cortex region. This applied not only in SDT rat diabetic nephropathy but also in puromycin aminonucleoside-induced nephropathy, which was also studied. The proposed glomerulus image analysis method combined with desmin immunohistochemistry should facilitate evaluations in preclinical drug efficacy studies as well as elucidation of the pathophysiology of diabetic nephropathy.
Product Description
Supelco
Losartan potassium
Losartan potassium, European Pharmacopoeia (EP) Reference Standard