Merck
  • Decatenation checkpoint-defective melanomas are dependent on PI3K for survival.
Decatenation checkpoint-defective melanomas are dependent on PI3K for survival.
Pigment cell & melanoma research (2014-06-04)
Kelly Brooks, Max Ranall, Loredana Spoerri, Alex Stevenson, Gency Gunasingh, Sandra Pavey, Fred Meunier, Thomas J Gonda, Brian Gabrielli
ABSTRACT
Melanoma cell lines are commonly defective for the G2-phase cell cycle checkpoint that responds to incomplete catenation of the replicated chromosomes. Here, we demonstrate that melanomas defective for this checkpoint response are less sensitive to genotoxic stress, suggesting that the defective cell lines compensated for the checkpoint loss by increasing their ability to cope with DNA damage. We performed an siRNA kinome screen to identify kinases responsible and identified PI3K pathway components. Checkpoint-defective cell lines were three-fold more sensitive to small molecule inhibitors of PI3K. The PI3K inhibitor PF-05212384 promoted apoptosis in the checkpoint-defective lines, and the increased sensitivity to PI3K inhibition correlated with increased levels of activated Akt. This work demonstrates that increased PI3K pathway activation is a necessary adaption for the continued viability of melanomas with a defective decatenation checkpoint.
Sigma-Aldrich
MISSION® esiRNA, targeting human POLR2A
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Polr2a
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
SAFC
HEPES
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
SAFC
HEPES