Merck
  • Synthesis and anti-hepatitis B virus activity of C4 amide-substituted isosteviol derivatives.
Synthesis and anti-hepatitis B virus activity of C4 amide-substituted isosteviol derivatives.
Bioorganic & medicinal chemistry (2015-01-21)
Tsurng-Juhn Huang, Cheng-Lin Yang, Yu-Cheng Kuo, Yi-Chih Chang, Li-Ming Yang, Bo-Hon Chou, Shwu-Jiuan Lin
ABSTRACT
A series of novel isosteviol derivatives having C4-amide substituents were synthesized in order to test for antiviral effects against the hepatitis B virus (HBV) in vitro. Among them, IN-4 [N-(propylcarbonyl)-4α-amino-19-nor-ent-16-ketobeyeran] (5) exhibited inhibitory activity against secretion of HBsAg and HBeAg as well as inhibition of HBV DNA replication. Therefore, the mechanism of its antiviral activity was further analyzed using HBV-transfected Huh7 cells. Exposure to IN-4 produced minimal inhibitory effects on viral precore/pregenomic RNA expression. However, expression levels of the 2.4/2.1-kb preS/major S RNA of the viral surface gene significantly decreased, along with intracellular levels of HBV DNA. A promoter activity analysis demonstrated that IN-4 significantly inhibited viral X, S, and preS expression levels but not viral core promoter activities. In particular, IN-4 was observed to significantly inhibit HBV gene regulation by disrupting nuclear factor (NF)-κB-associated promoter activity. In addition, the nuclear expression of p65/p50 NF-κB member proteins was attenuated following IN-4 treatment, while cytoplasmic IκBα protein levels were enhanced. Meanwhile, IN-4 was observed to inhibit the binding activity of NF-κB to putative DNA elements. Furthermore, transfection of a p65 expression plasmid into Huh7 cells significantly reversed the inhibitory effect of IN-4 on HBV DNA levels, providing further evidence of the central role of NF-κB in its antiviral mechanism. It is therefore suggested that IN-4 inhibits HBV by interfering with the NF-κB signaling pathway, resulting in downregulation of viral gene expression and DNA replication.
Product Description
Lamivudine for system suitability 2, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Supelco
Lamivudine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lamivudine, ≥98% (HPLC), powder
Lamivudine, European Pharmacopoeia (EP) Reference Standard
Lamivudine for system suitability 1, European Pharmacopoeia (EP) Reference Standard