Merck
  • Anti-metastatic immunotherapy based on mucosal administration of flagellin and immunomodulatory P10.
Anti-metastatic immunotherapy based on mucosal administration of flagellin and immunomodulatory P10.
Immunology and cell biology (2014-09-17)
Filipe M de Melo, Catarina J M Braga, Felipe V Pereira, Juliana T Maricato, Clarice S T Origassa, Mariana F Souza, Amanda C Melo, Priscila Silva, Samanta L Tomaz, Karina P Gimenes, Jorge A B Scutti, Maria A Juliano, Dario S Zamboni, Niels O Câmara, Luiz R Travassos, Luis C S Ferreira, Elaine G Rodrigues
ABSTRACT
Current therapies against malignant melanoma generally fail to increase survival in most patients, and immunotherapy is a promising approach as it could reduce the dosage of toxic therapeutic drugs. In the present study, we show that an immunotherapeutic approach based on the use of the Toll-like receptor (TLR)-5 ligand flagellin (Salmonella Typhimurium FliCi) combined with the major histocompatibility complex class II-restricted P10 peptide, derived from the Paracoccidioides brasiliensis gp43 major surface protein, reduced the number of lung metastasis in a murine melanoma model. Compounds were administered intranasally into C57Bl/6 mice intravenously challenged with syngeneic B16F10-Nex2 melanoma cells, aiming at the local (pulmonary) immune response modulation. Along with a marked reduction in the number of lung nodules, a significant increase in survival was observed. The immunization regimen induced both local and systemic proinflammatory responses. Lung macrophages were polarized towards a M1 phenotype, lymph node cells, and splenocytes secreted higher interleukin-12p40 and interferon (IFN)-γ levels when re-stimulated with tumor antigens. The protective effect of the FliCi+P10 formulation required TLR-5, myeloid differentiation primary response gene 88 and IFN-γ expression, but caspase-1 knockout mice were only partially protected, suggesting that intracellular flagellin receptors are not involved with the anti-tumor effect. The immune therapy resulted in the activation of tumor-specific CD4(+) T lymphocytes, which conferred protection to metastatic melanoma growth after adoptive transfer. Taken together, our results report a new immunotherapeutic approach based on TLR-5 activation and IFN-γ production capable to control the metastatic growth of B16F10-Nex2 melanoma, being a promising alternative to be associated with chemotherapeutic drugs for an effective anti-tumor responses.
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
SAFC
HEPES
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%