Merck
  • Separation of porcine parvovirus from bovine serum albumin using PEG-salt aqueous two-phase system.
Separation of porcine parvovirus from bovine serum albumin using PEG-salt aqueous two-phase system.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-08-03)
K Saagar Vijayaragavan, Amna Zahid, Jonathan W Young, Caryn L Heldt
ABSTRACT
Vaccine production faces a challenge in adopting conventional downstream processing steps that can efficiently purify large viral particles. Some major issues that plague vaccine purification are purity, potency, and quality. The industry currently considers 30% as an acceptable virus recovery for a vaccine purification process, including all downstream processes, whereas antibody recovery from CHO cell culture is generally around 80-85%. A platform technology with an improved virus recovery would revolutionize vaccine production. In a quest to fulfill this goal, we have been exploring aqueous two-phase systems (ATPSs) as an optional mechanism to purify virus. ATPS has been unable to gain wide implementation mainly due to loss of virus infectivity, co-purification of proteins, and difficulty of polymer recycling. Non-enveloped viruses are chemically resistant enough to withstand the high polymer and salt concentrations that are required for effective ATPS separations. We used infectious porcine parvovirus (PPV), a non-enveloped, DNA virus as a model virus to test and develop an ATPS separation method. We successfully tackled two of the three main disadvantages of ATPS previously stated; we achieved a high infectious yield of 64% in a PEG-citrate ATPS process while separating out the main contaminate protein, bovine serum albumin (BSA). The most dominant forces in the separation were biomolecule charge, virus surface hydrophobicity, and the ATPS surface tension. Highly hydrophobic viruses are likely to benefit from the discovered ATPS for high-purity vaccine production and ease of implementation.
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Trisodium citrate dihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)