Merck
  • In vitro and in vivo activity of the low-immunogenic antimesothelin immunotoxin RG7787 in pancreatic cancer.
In vitro and in vivo activity of the low-immunogenic antimesothelin immunotoxin RG7787 in pancreatic cancer.
Molecular cancer therapeutics (2014-06-15)
Kevin Hollevoet, Emily Mason-Osann, Xiu-fen Liu, Sabine Imhof-Jung, Gerhard Niederfellner, Ira Pastan
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) has a dismal prognosis, and new therapies are needed. RG7787 is a novel low-immunogenic antimesothelin recombinant immunotoxin (RIT), engineered to overcome the limitations of SS1P, a RIT now in clinical trials. In vitro activity was evaluated on five established PDAC cell lines (KLM-1, AsPC-1, BxPC-3, Panc 3.014, and PK-1) and on PDAC cells directly established from a patient tumor (GUMC108). RG7787 had subnanomolar IC50s in most cell lines, and was significantly more active than SS1P in GUMC108, KLM-1, and Panc 3.014 cells. GUMC108 was most sensitive, with RG7787 killing >99% of the cells. In a subcutaneous KLM-1 xenograft mouse model, two cycles of 3 × 2.5 mg/kg RG7787 QOD combined with two cycles of 1 × 50 mg/kg paclitaxel induced near-complete responses, with all tumors regressing below 5 mm(3) within 30 days after therapy was initiated (>95% decrease) and no significant growth increase for at least another 3 weeks. RG7787 alone gave limited but significant regressions and paclitaxel by itself arrested tumor growth. Quantifying the uptake of Alexa Fluor 647-labeled RG7787 in tumors showed that the RIT reached only 45% of KLM-1 cells, accounting in part for the limited responses. Paclitaxel did not improve RG7787 uptake, which thus cannot explain the beneficial effect of the combination therapy. In conclusion, RG7787 has high cytotoxic activity on PDAC cell lines as well as on primary patient cells. In vivo, this novel RIT gives durable near-complete tumor responses when combined with paclitaxel. RG7787 merits further evaluation for the treatment of PDAC.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Staurosporine, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Adenine, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Pricing and availability is not currently available.
Adenine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Pricing and availability is not currently available.
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Pricing and availability is not currently available.
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.