Merck
  • A6V polymorphism of the human μ-opioid receptor decreases signalling of morphine and endogenous opioids in vitro.
A6V polymorphism of the human μ-opioid receptor decreases signalling of morphine and endogenous opioids in vitro.
British journal of pharmacology (2014-12-19)
Alisa Knapman, Marina Santiago, Mark Connor
ABSTRACT
Polymorphisms of the μ opioid receptor (MOPr) may contribute to the variation in responses to opioid drugs in clinical and unregulated situations. The A6V variant of MOPr (MOPr-A6V) is present in up to 20% of individuals in some populations, and may be associated with heightened susceptibility to drug abuse. There are no functional studies examining the acute signalling of MOPr-A6V in vitro, so we investigated potential functional differences between MOPr and MOPr-A6V at several signalling pathways using structurally distinct opioid ligands. CHO and AtT-20 cells stably expressing MOPr and MOPr-A6V were used. AC inhibition and ERK1/2 phosphorylation were assayed in CHO cells; K channel activation was assayed in AtT-20 cells. Buprenorphine did not inhibit AC or stimulate ERK1/2 phosphorylation in CHO cells expressing MOPr-A6V, but buprenorphine activation of K channels in AtT-20 cells was preserved. [D-Ala2, N-MePhe4, Gly-ol]-enkephalin, morphine and β-endorphin inhibition of AC was significantly reduced via MOPr-A6V, as was signalling of all opioids to ERK1/2. However, there was little effect of the A6V variant on K channel activation. Signalling to AC and ERK via the mutant MOPr-A6V was decreased for many opioids, including the clinically significant drugs morphine, buprenorphine and fentanyl, as well endogenous opioids. The MOPr-A6V variant is common and this compromised signalling may affect individual responses to opioid therapy, while the possible disruption of the endogenous opioid system may contribute to susceptibility to substance abuse.
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Supelco
Forskolin, analytical standard
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)