Merck
  • A novel zinc finger gene, ZNF465, is inappropriately expressed in acute myeloid leukaemia cells.
A novel zinc finger gene, ZNF465, is inappropriately expressed in acute myeloid leukaemia cells.
Genes, chromosomes & cancer (2015-02-24)
Joseph F Collin, James W Wells, Barbara Czepulkowski, Linden Lyne, Patrick J Duriez, Alison H Banham, Ghulam J Mufti, Barbara-Ann Guinn
ABSTRACT
To increase our knowledge of leukaemia-associated antigens, especially in acute myeloid leukaemia (AML) M4, we prepared a phage display cDNA library using mRNA from the bone marrow cells of a patient with AML M4 at diagnosis. We immunoscreened 10(6) pfu with autologous sera and identified an antigen which we named GKT-AML8. The cDNA showed more than 99% similarity to a sequence on 2q21.2 and 95% sequence similarity to a sequence on 19q13.3. These genes were named ZNF465 and ZNF466, respectively, following HUGO Gene Nomenclature Committee (HGNC) guidelines. Expressed sequence tag data suggests that both genes are transcriptionally active. ZNF465 and ZNF466 encode a 5' krüppel associated box domain typical of negative regulators of gene transcription. We have confirmed the translational start site in the +1 frame in a near-Kozak sequence that produces a 102 amino acid polypeptide from ZNF465. The high level of sequence similarity between ZNF465 and ZNF466 makes their transcripts almost indistinguishable by real-time polymerase chain reaction (RT-PCR). However, GKT-AML8 showed most sequence similarity to ZNF465 and no transcript matching the 3' ZNF466 sequence could be detected in patient samples or healthy volunteers. ZNF465/466 expression was detectable in 12/13 AML and 10/14 chronic myeloid leukaemia patients' samples but not in normal donor peripheral blood (0/8) or 0/3 bone marrow samples which had been separated into CD34(+) and CD34(-) samples. The altered expression of ZNF465/466 in patients' samples and its absence in healthy donor haematopoietic samples indicate that ZNF465 is overexpressed in early myeloid disease and as such may represent a promising target for immunotherapy.
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture