Merck
  • Nasopharyngeal carriage of Streptococcus pneumoniae in Romanian children before the introduction of the pneumococcal conjugated vaccination into the national immunization programme: a national, multi-centre, cross-sectional observational study.
Nasopharyngeal carriage of Streptococcus pneumoniae in Romanian children before the introduction of the pneumococcal conjugated vaccination into the national immunization programme: a national, multi-centre, cross-sectional observational study.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases (2014-12-03)
Monica Luminos, Olga Dorobat, George Jugulete, Gabriel Adrian Popescu, Dragos Florea, Anca Draganescu, Anca Streinu Cercel, Alexandru Rafila
ABSTRACT
We analysed the distribution of vaccine and non-vaccine Streptococcus pneumoniae serotypes and the antimicrobial susceptibility of pneumococcal strains isolated from healthy Romanian children. A multi-centre cross-sectional study was performed in four counties to evaluate carried strains of S. pneumoniae isolated from 2000 children aged 0-5 years. S. pneumoniae carriage was detected in 25.25% of the tested children. Carriage increased from 16.7% among infants to 29.4% in 3-5-year-old children (p<0.0001). The proportions of the serotypes included in pneumococcal conjugate vaccines PCV7, PCV10, and PCV13 among our isolates were 39.9%, 40.1%, and 58.7%, respectively. Erythromycin resistance was 72.5%, and it was significantly lower in non-vaccine serotypes compared with PCV13 serotypes: 57.3% versus 83.6% (p<10(-7)). Penicillin minimum inhibitory concentrations (MICs) >0.064mg/l were recorded in 71.6%, but the penicillin MIC was >2mg/l for only 8.4% of tested isolates. In Romanian children, the majority of carried S. pneumoniae isolates are vaccine serotypes. The isolates with MICs defining macrolide resistance were very frequent, as well as the isolates with MICs defining penicillin resistance in the case of meningitis or penicillin dose-dependent susceptibility for other infections, mainly for the strains belonging to PCV13 serotypes. The implementation of PCV13 within the Romanian national immunization programme could reduce the circulation of these strains with higher macrolide and/or penicillin MICs.
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard