Merck
  • SAFB1- and SAFB2-mediated transcriptional repression: relevance to cancer.
SAFB1- and SAFB2-mediated transcriptional repression: relevance to cancer.
Biochemical Society transactions (2012-07-24)
Elaine A Hong, Hannah L Gautrey, David J Elliott, Alison J Tyson-Capper
ABSTRACT
SAFB1 (scaffold attachment factor B1) and a second family member SAFB2, are multifunctional proteins implicated in a variety of cellular processes including cell growth, apoptosis and stress response. Their potential function as tumour suppressors has been proposed based on well-described roles in tran-scriptional repression. The present review summarizes the current knowledge of SAFB1 and SAFB2 proteins in transcriptional repression with relevance to cancer.