Merck
  • Lower expression of Nrdp1 in human glioma contributes tumor progression by reducing apoptosis.
Lower expression of Nrdp1 in human glioma contributes tumor progression by reducing apoptosis.
IUBMB life (2014-10-31)
Hengliang Shi, Jin Du, Lei Wang, Bao Zheng, Hui Gong, Yuxuan Wu, Yuan Tang, Yong Gao, Rutong Yu
ABSTRACT
Ubiquitin ligase Nrdp1 (neuregulin receptor degradation protein 1) plays important roles in multiple physiological process because it can ubiquitinate various substrates such as ErbB3, BRUCE, MyD88, C/EBPβ, and Parkin, and so forth. In addition to the physiological function, it was also found to be involved in tumor progression. It has been shown that loss of Nrdp1 enhances breast cancer cell growth. Up to now, the role of Nrdp1 in glioma has not been elucidated. Here, we reported that Nrdp1 as well as cleaved caspase 3 was lower expressed in human glioma tissues comparing with the nontumorous. And then we found that the expression of Nrdp1 and cleaved caspase 3 was increased in the treatment of Temozolomide (TMZ), a drug for glioma chemotherapy. Further investigation indicated that transient transfection of Nrdp1 significantly promoted cell apoptosis by aggravating the degradation of BRUCE and activation of caspase 3. In addition, overexpression of Nrdp1 augmented TMZ induced apoptosis by evaluating the degradation of BRUCE and the activation of caspase 3, while silencing of Nrdp1 reduced the sensitivity to the TMZ by inhibiting the degradation of BRUCE and the activation of caspase 3 in human glioma cells. These observations show that Nrdp1 is a pro-apoptotic protein in human glioma and lower expression of Nrdp1 in human glioma may promote tumor progression by reducing apoptosis, suggesting that Nrdp1 may be an important regulator in the development of human glioma.