Merck
  • Organ weaving: woven threads and sheets as a step towards a new strategy for artificial organ development.
Organ weaving: woven threads and sheets as a step towards a new strategy for artificial organ development.
Macromolecular bioscience (2011-09-15)
Albert R Liberski, Joseph T Delaney, Hendrik Schäfer, Jolke Perelaer, Ulrich S Schubert
ABSTRACT
The concept of "organ weaving" is presented, a fabrication technique that can be an attractive option for the development of artificial tissues and organs. "Living threads" are created by immersing threads that are soaked in a CaCl(2) solution into a sodium-alginate-loaded cell suspension bath, encapsulating the cells and creating a bio-friendly, easily manageable starting material for building up larger scaffold structures. Such living threads have the advantage of being a particularly mild culturing medium for mammalian cells, protecting the cells during subsequent processing steps from dehydration and other rapid changes in the chemistry of the surrounding environment. Connecting different types of threads into 3D objects gives unique opportunities to address tissue engineering challenges.
Product Description
Greiner centrifuge tubes, centrifuge tube, 50 mL, 30 x 115 mm, conical (V) bottom, w/ graduations, I.D. field
Pricing and availability is not currently available.