Merck
  • Antimycobacterial and HIV-1 Reverse Transcriptase Activity of Julianaceae and Clusiaceae Plant Species from Mexico.
Antimycobacterial and HIV-1 Reverse Transcriptase Activity of Julianaceae and Clusiaceae Plant Species from Mexico.
Evidence-based complementary and alternative medicine : eCAM (2015-05-20)
Rocio Gómez-Cansino, Clara Inés Espitia-Pinzón, María Guadalupe Campos-Lara, Silvia Laura Guzmán-Gutiérrez, Erika Segura-Salinas, Gabriela Echeverría-Valencia, Laura Torras-Claveria, Xochitl Marisol Cuevas-Figueroa, Ricardo Reyes-Chilpa
ABSTRACT
The extracts of 14 Julianaceae and 5 Clusiaceae species growing in Mexico were tested in vitro (50 µg/mL) against Mycobacterium tuberculosis H37Rv and HIV reverse transcriptase (HIV-RT). The Julianaceae bark and leaf extracts inhibited M. tuberculosis (>84.67%) and HIV-RT (<49.89%). The Clusiaceae leaves extracts also inhibited both targets (>58.3% and >67.6%), respectively. The IC50 values for six selected extracts and their cytotoxicity (50 µg/mL) to human macrophages were then determined. Amphipterygium glaucum, A. molle, and A. simplicifolium fairly inhibited M. tuberculosis with IC50 of 1.87-2.35 µg/mL; but their IC50 against HIV-RT was 59.25-97.83 µg/mL. Calophyllum brasiliense, Vismia baccifera, and Vismia mexicana effect on M. tuberculosis was noteworthy (IC50 3.02-3.64 µg/mL) and also inhibited RT-HIV (IC50 26.24-35.17 µg/mL). These 6 extracts (50 µg/mL) presented low toxicity to macrophages (<23.8%). The HPLC profiles of A. glaucum, A. molle, and A. simplicifolium indicated that their antimycobacterial activity cannot be related to masticadienonic, 3α, or 3β-hydromasticadienonic acids, suggesting that other compounds may be responsible for the observed activity or this might be a synergy result. The anti-HIV-RT and antimycobacterial activities induced by C. brasiliense can be attributed to the content of calanolides A, B, as well as soulatrolide.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Pricing and availability is not currently available.