Merck
  • An expeditious synthesis of N-substituted pyrroles via microwave-induced iodine-catalyzed reactions under solventless conditions.
An expeditious synthesis of N-substituted pyrroles via microwave-induced iodine-catalyzed reactions under solventless conditions.
Molecules (Basel, Switzerland) (2010-04-30)
Debasish Bandyopadhyay, Sanghamitra Mukherjee, Bimal K Banik
ABSTRACT
An expeditious synthesis of N-substituted pyrroles has been developed by reacting 2,5-dimethoxy tetrahydrofuran and several amines using a microwave-induced molecular iodine-catalyzed reaction under solventless conditions.