- Enhanced function of redirected human T cells expressing linker for activation of T cells that is resistant to ubiquitylation.
Enhanced function of redirected human T cells expressing linker for activation of T cells that is resistant to ubiquitylation.
It is likely that the enhancement of signaling after antigenic stimulation, particularly in the tumor microenvironment, would improve the function of adoptively transferred T cells. Linker for activation of T cells (LAT) plays a central role in T cell activation. We hypothesized that the ubiquitylation-resistant form of LAT in cells would enhance T cell signaling and thus augment antitumor activity. To test this, human CD4(+) or CD8(+) T cells were electroporated with small interfering RNA (siRNA) to repress endogenous LAT and ubiquitylation-resistant LAT 2KR or wild-type LAT mRNA was introduced for reexpression. Significantly enhanced phosphorylation of LAT and phospholipase C-γ (PLCγ) was observed, and augmented calcium signaling after T cell receptor (TCR) triggering was observed in LAT 2KR-expressing T cells. TCR-induced calcium signaling was abrogated in LAT knockdown cells, but the baseline was higher than that of control siRNA-electroporated cells, suggesting a fundamental requirement of LAT to maintain calcium homeostasis. Redirected LAT 2KR T cells expressing a chimeric antigen receptor or an MHC class I-restricted TCR showed augmented function as assessed by enhanced cytokine secretion and cytotoxicity. These results indicate that interruption of LAT ubiquitylation is a promising strategy to augment effector T cell function for adoptive cell therapy.