Merck
  • Alcoholic liver disease and the mitochondrial ribosome: methods of analysis.
Alcoholic liver disease and the mitochondrial ribosome: methods of analysis.
Methods in molecular biology (Clifton, N.J.) (2008-03-29)
Alan Cahill, Peter Sykora
ABSTRACT
Chronic alcohol consumption has been shown to severely compromise mitochondrial protein synthesis. Hepatic mitochondria isolated from alcoholic animals contain decreased levels of respiratory complexes and display depressed respiration rates when compared to pair-fed controls. One underlying mechanism for this involves ethanol-elicited alterations in the structural and functional integrity of the mitochondrial ribosome. Ethanol feeding results in ribosomal changes that include decreased sedimentation rates, larger hydrodynamic volumes, increased levels of unassociated subunits and changes in the levels of specific ribosomal proteins. The methods presented in this chapter detail how to isolate mitochondrial ribosomes, determine ribosomal activity, separate ribosomes into nucleic acid and protein, and perform two-dimensional nonequilibrium pH gradient electrophoretic polyacrylamide gel electrophoresis to separate and subsequently identify mitochondrial ribosomal proteins.
Sigma-Aldrich
Phospho(enol)pyruvic acid monopotassium salt, ≥97% (enzymatic)
Pricing and availability is not currently available.