Merck
  • Composition of essential oils isolated from the needles of Pinus uncinata and P. uliginosa grown in Poland.
Composition of essential oils isolated from the needles of Pinus uncinata and P. uliginosa grown in Poland.
Natural product communications (2015-04-30)
Radosław Bonikowski, Konrad Celiński, Aleksandra Wojnicka-Półtorak, Tomasz Maliński
ABSTRACT
The compositions of mountain pine (Pinus uncinata) and peat-bog pine (P. uliginosa) needle essential oils were investigated. Enantiomeric compositions of selected monoterpene hydrocarbons were also examined. Respectively, fifty-three and seventy-six components of the essential oils were identified using GC-MS and retention indexes. The main group of essential oil components of mountain pine needles were monoterpenes, and bornyl acetate constituted approximately 30% (46.3 g/100 g) of the oil. In peat-bog pine essential oil, monoterpenes and sesquiterpenes exhibited a similar content (ca. 40%). Bornyl acetate and α-pinene were the main constituents of both essential oils. In the essential oil of P. uncinata needles, limonene, camphene, myrcene and (E)-β-caryophyllene were also noticeable, while in the essential oil of P. uliginosa needles, Δ-car-3-ene, (E)-β-caryophyllene, germacrene D, δ-cadinene, germacrene D 4-ol and α-cadinol were present in notable quantities. In both essential oils, borneol propionate, isobutyrate, 2-methylbutyrate and isovalerate were detected. Their presence was confirmed by synthesis and analysis of the standards; retention indexes on a non-polar column are published herein.
Sigma-Aldrich
Camphene, 95%
Pricing and availability is not currently available.